siRNA Details
Gene Symbol: SDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035511.1 | NP_001030588.1 | 2, 3 | 109 | ||
NM_003001.3 | NP_002992.1 | 2, 3 | 109 | |||
NR_103459.1 | 2, 3 | 109 | ||||
GenBank mRNA | AB211235.1 | - | 2, 3 | 103 | ||
AK131051.1 | - | 2, 3 | 104 | |||
AK294305.1 | - | 2, 3 | 104 | |||
BC020808.1 | - | 2, 3 | 104 | 4291847 | ||
BC033626.1 | - | 2, 3 | 102 | 5416956 | ||
BC066329.1 | - | 2, 3 | 104 | 5301907 | ||
BG724095.1 | - | 2, 3 | 233 | |||
CR457102.1 | - | 1, 2 | 79 | |||
D49737.1 | - | 2, 3 | 92 | |||
DA751501.1 | - | 2, 3 | 94 | |||
KU178321.1 | - | 2, 3 | 79 | |||
KU178323.1 | - | 2, 3 | 79 | |||
U57877.1 | - | 2, 3 | 106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
