siRNA Details
Gene Symbol: SDHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035511.1 | NP_001030588.1 | 5 | 714 | ||
NM_001035512.1 | NP_001030589.1 | 5 | 776 | |||
NM_001035513.1 | NP_001030590.1 | 4 | 719 | |||
NM_001278172.1 | NP_001265101.1 | 4 | 612 | |||
NM_003001.3 | NP_002992.1 | 6 | 878 | |||
NR_103459.1 | 7 | 905 | ||||
GenBank mRNA | AK131051.1 | - | 6 | 873 | ||
AK293687.1 | - | 5 | 768 | |||
AK294305.1 | - | 5 | 709 | |||
BC020808.1 | - | 6 | 873 | 4291847 | ||
BC033626.1 | - | 6 | 871 | 5416956 | ||
BC066329.1 | - | 6 | 873 | 5301907 | ||
D49737.1 | - | 6 | 861 | |||
U57877.1 | - | 6 | 875 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
