siRNA Details
Gene Symbol: PRIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000947.4 | NP_000938.2 | 4, 5 | 432 | ||
NM_001282487.1 | NP_001269416.1 | 4, 5 | 432 | |||
NM_001282488.1 | NP_001269417.1 | 4, 5 | 463 | |||
XM_017011004.1 | XP_016866493.1 | 4, 5 | 432 | |||
XM_017011005.1 | XP_016866494.1 | 4, 5 | 432 | |||
XM_017011006.1 | XP_016866495.1 | 4, 5 | 432 | |||
XM_017011007.1 | XP_016866496.1 | 3, 4 | 347 | |||
GenBank mRNA | AK223287.1 | - | 4, 5 | 421 | ||
AK291915.1 | - | 4, 5 | 419 | |||
BC017833.1 | - | 4, 5 | 432 | |||
BC064931.1 | - | 4, 5 | 417 | 6148494 | ||
BI547222.1 | - | 4, 5 | 468 | |||
X74331.1 | - | 4, 5 | 425 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
