siRNA Details
Gene Symbol: PDSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321978.1 | NP_001308907.1 | 3 | 467 | ||
NM_001321979.1 | NP_001308908.1 | 3 | 467 | |||
NM_014317.4 | NP_055132.2 | 3 | 467 | |||
XM_017016011.1 | XP_016871500.1 | 2 | 54 | |||
GenBank mRNA | AB209763.1 | - | 2 | 2505 | ||
AB210838.1 | - | 3 | 225 | |||
AK024802.1 | - | 3 | 232 | |||
AK223414.1 | - | 3 | 232 | |||
BC049211.1 | - | 3 | 177 | |||
BC063635.1 | - | 3 | 195 | 4397753 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
