siRNA Details
Gene Symbol: NPFFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022146.4 | NP_071429.1 | 4 | 994 | ||
GenBank mRNA | AB040104.1 | - | 3 | 976 | ||
AF268898.1 | - | 3 | 976 | |||
AF330055.1 | - | 3 | 976 | |||
BC131580.1 | - | 3 | 976 | 40112416 | ||
EU432128.1 | - | 3 | 976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
