siRNA Details
Gene Symbol: TRIM48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024114.3 | NP_077019.2 | 2 | 327 | ||
GenBank mRNA | AF521869.1 | - | 2 | 334 | ||
BC001862.2 | - | 2 | 327 | 3603763 |
Gene Symbol: TRIM49D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206627.1 | NP_001193556.1 | 1 | 193 | ||
XM_011542816.2 | XP_011541118.1 | 2 | 309 |
Gene Symbol: TRIM49D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105522.1 | NP_001098992.1 | 2 | 307 |
Gene Symbol: TRIM51HP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB231736.1 | - | 1 | 156 |
Gene Symbol: TRIM51JP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | JF968455.1 | - | 1 | 115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
