siRNA Details
Gene Symbol: USP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182488.3 | NP_872294.2 | 3 | 475 | ||
XM_005266282.2 | XP_005266339.1 | 3 | 389 | |||
XM_011534956.2 | XP_011533258.1 | 2 | 143 | |||
GenBank mRNA | AF022789.1 | - | 3 | 325 | ||
AK289685.1 | - | 3 | 389 | |||
BC026072.1 | - | 3 | 476 | 4830251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
