siRNA Details
Gene Symbol: TECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321170.1 | NP_001308099.1 | 9, 10 | 764 | ||
NM_138501.5 | NP_612510.1 | 8, 9 | 690 | |||
NR_038103.1 | 8, 9 | 722 | ||||
NR_038104.1 | 8, 9 | 755 | ||||
XM_006722945.1 | XP_006723008.1 | 9, 10 | 808 | |||
XM_011528442.2 | XP_011526744.1 | 8, 9 | 638 | |||
GenBank mRNA | AF038958.1 | - | 8, 9 | 636 | ||
AF038959.1 | - | 3, 4 | 119 | |||
AF222742.1 | - | 8, 9 | 611 | |||
AK001416.1 | - | 8, 9 | 654 | |||
AK094087.1 | - | 9, 10 | 731 | |||
AK299134.1 | - | 9, 10 | 728 | |||
AK315412.1 | - | 8, 9 | 654 | |||
BC000174.2 | - | 7, 8 | 550 | |||
BC002425.1 | - | 8, 9 | 622 | 3163592 | ||
BC005952.1 | - | 8, 9 | 633 | 4249841 | ||
BC007801.2 | - | 8, 9 | 640 | 4128702 | ||
BC013881.2 | - | 8, 9 | 638 | 3928698 | ||
BT007179.1 | - | 7, 8 | 560 | |||
CR456892.1 | - | 7, 8 | 560 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
