siRNA Details
Gene Symbol: MIA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324062.1 | NP_001310991.1 | 4 | 2327 | ||
NM_001324063.1 | NP_001310992.1 | 4 | 2327 | |||
NM_001324064.1 | NP_001310993.1 | 5 | 2372 | |||
NM_198551.3 | NP_940953.2 | 4 | 2327 | |||
XM_006711304.3 | XP_006711367.2 | 4 | 2639 | |||
XM_011509513.2 | XP_011507815.2 | 4 | 2639 | |||
XM_017001243.1 | XP_016856732.1 | 4 | 2152 | |||
GenBank mRNA | AK096526.1 | - | 1 | 1065 | ||
AK307412.1 | - | 4 | 2323 | |||
D87742.1 | - | 1 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
