siRNA Details
Gene Symbol: CCDC144A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014695.2 | NP_055510.1 | 3 | 512 | ||
NR_130142.1 | 3 | 512 | ||||
XM_011524089.2 | XP_011522391.1 | 3 | 602 | |||
XM_017025429.1 | XP_016880918.1 | 3 | 529 | |||
XM_017025430.1 | XP_016880919.1 | 3 | 602 | |||
GenBank mRNA | BC133019.1 | - | 3 | 512 | 40147041 | |
BC144224.1 | - | 3 | 512 | 9052744 | ||
BC144225.1 | - | 3 | 512 | 9052745 |
Gene Symbol: CCDC144B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036647.1 | 3 | 606 | |||
GenBank mRNA | AK093811.1 | - | 3 | 606 | ||
BC062767.1 | - | 3 | 600 | |||
BC101585.1 | - | 3 | 521 | 8069091 | ||
BC104815.1 | - | 3 | 521 | 8143818 | ||
BC143561.1 | - | 3 | 521 | 9052072 | ||
BX647826.1 | - | 3 | 597 |
Gene Symbol: CCDC144CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023380.1 | 3 | 579 | |||
GenBank mRNA | AL162090.1 | - | 3 | 579 | ||
BC036241.1 | - | 3 | 591 |
Gene Symbol: LOC101929141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025498.1 | XP_016880987.1 | 1 | 64 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
