siRNA Details
Gene Symbol: LOC101929141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017025498.1 | XP_016880987.1 | 1, 2 | 290 |
Gene Symbol: CCDC144A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014695.2 | NP_055510.1 | 3, 4 | 738 | ||
NR_130142.1 | 3, 4 | 738 | ||||
XM_011524089.2 | XP_011522391.1 | 3, 4 | 828 | |||
XM_017025429.1 | XP_016880918.1 | 3, 4 | 755 | |||
XM_017025430.1 | XP_016880919.1 | 3, 4 | 828 | |||
GenBank mRNA | BC133019.1 | - | 3, 4 | 738 | 40147041 | |
BC144224.1 | - | 3, 4 | 738 | 9052744 | ||
BC144225.1 | - | 3, 4 | 738 | 9052745 |
Gene Symbol: CCDC144B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036647.1 | 3, 4 | 832 | |||
GenBank mRNA | BC062767.1 | - | 3, 4 | 826 | ||
BC101585.1 | - | 3, 4 | 747 | 8069091 | ||
BC104815.1 | - | 3, 4 | 747 | 8143818 | ||
BX647826.1 | - | 3, 4 | 823 |
Gene Symbol: CCDC144CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023380.1 | 3, 4 | 805 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
