siRNA Details
Gene Symbol: TBC1D28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039397.2 | NP_001034486.2 | 5 | 537 | ||
XM_011523781.2 | XP_011522083.1 | 5 | 537 | |||
XM_011523782.2 | XP_011522084.1 | 5 | 417 | |||
XM_011523783.2 | XP_011522085.1 | 4 | 445 | |||
XM_017024416.1 | XP_016879905.1 | 4 | 449 | |||
XM_017024417.1 | XP_016879906.1 | 5 | 496 | |||
XM_017024418.1 | XP_016879907.1 | 5 | 594 | |||
XM_017024419.1 | XP_016879908.1 | 4 | 420 | |||
XM_017024420.1 | XP_016879909.1 | 5 | 1888 | |||
XM_017024421.1 | XP_016879910.1 | 5 | 467 | |||
XM_017024422.1 | XP_016879911.1 | 4 | 448 | |||
XM_017024423.1 | XP_016879912.1 | 4 | 465 | |||
GenBank mRNA | AK097563.1 | - | 4 | 825 | ||
BC112012.1 | - | 3 | 203 | 8327480 | ||
BC112016.1 | - | 3 | 203 | 8327484 |
Gene Symbol: TBC1D26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178571.4 | NP_848666.2 | 4 | 375 | ||
GenBank mRNA | AB449913.1 | - | 2 | 125 | ||
AK292544.1 | - | 4 | 370 | |||
AK315847.1 | - | 4 | 370 | |||
BC028414.1 | - | 4 | 374 | |||
BC047400.1 | - | 4 | 369 | 5297645 | ||
DB067071.1 | - | 4 | 413 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
