siRNA Details
Gene Symbol: TMEM171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161342.2 | NP_001154814.1 | 2 | 758 | ||
NM_173490.7 | NP_775761.4 | 2 | 758 | |||
XM_011543156.1 | XP_011541458.1 | 2 | 738 | |||
GenBank mRNA | BC018083.1 | - | 2 | 744 | 4797244 | |
BC035310.1 | - | 1 | 599 | 4746772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
