siRNA Details
Gene Symbol: TMEM171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173490.7 | NP_775761.4 | 3, 4 | 979 | ||
XM_011543156.1 | XP_011541458.1 | 3, 4 | 902 | |||
GenBank mRNA | BC018083.1 | - | 3, 4 | 965 | 4797244 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
