siRNA Details
Gene Symbol: C1R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001733.4 | NP_001724.3 | 11 | 1501 | ||
XM_017019915.1 | XP_016875404.1 | 11 | 1575 | |||
GenBank mRNA | AK024951.1 | - | 9 | 2507 | ||
AK094009.1 | - | 4 | 1791 | |||
AK222481.1 | - | 11 | 1495 | |||
AK222491.1 | - | 11 | 1495 | |||
AK291313.1 | - | 11 | 1523 | |||
AK298445.1 | - | 11 | 1516 | |||
AK303750.1 | - | 11 | 1394 | |||
BC035220.1 | - | 11 | 1501 | 4777940 | ||
M14058.1 | - | 11 | 1505 | |||
X04701.1 | - | 11 | 1493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
