siRNA Details
Gene Symbol: MT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005946.2 | NP_005937.2 | 2 | 122 | ||
GenBank mRNA | AF348995.1 | - | 2 | 104 | ||
AY028617.1 | - | 2 | 122 | |||
BC029475.1 | - | 2 | 119 | 4723818 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
