siRNA Details
Gene Symbol: ANGEL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300753.1 | NP_001287682.1 | 3 | 563 | ||
NM_001300755.1 | NP_001287684.1 | 3 | 661 | |||
NM_001300757.1 | NP_001287686.1 | 2 | 316 | |||
NM_001300758.1 | NP_001287687.1 | 2 | 312 | |||
NM_144567.4 | NP_653168.2 | 3 | 638 | |||
NR_125333.1 | 2 | 312 | ||||
XM_005273344.1 | XP_005273401.1 | 3 | 659 | |||
XM_005273345.1 | XP_005273402.1 | 3 | 705 | |||
XM_005273346.2 | XP_005273403.1 | 3 | 458 | |||
XM_005273347.3 | XP_005273404.1 | 2 | 330 | |||
XM_017002774.1 | XP_016858263.1 | 3 | 587 | |||
XM_017002775.1 | XP_016858264.1 | 4 | 886 | |||
XM_017002776.1 | XP_016858265.1 | 4 | 910 | |||
XM_017002777.1 | XP_016858266.1 | 4 | 956 | |||
XM_017002778.1 | XP_016858267.1 | 4 | 699 | |||
GenBank mRNA | AF510741.1 | - | 2 | 267 | ||
AK294479.1 | - | 3 | 437 | |||
AK295116.1 | - | 3 | 567 | |||
AK299208.1 | - | 3 | 630 | |||
AK299365.1 | - | 2 | 222 | |||
AK311276.1 | - | 2 | 221 | |||
AL079275.1 | - | 3 | 593 | |||
AY468376.1 | - | 2 | 368 | |||
BC016966.1 | - | 2 | 382 | |||
BC047469.1 | - | 2 | 228 | 5274835 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
