siRNA Details
Gene Symbol: CAPN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144575.2 | NP_653176.2 | 2 | 222 | ||
XM_011533159.2 | XP_011531461.1 | 2 | 219 | |||
XM_011533160.2 | XP_011531462.1 | 3 | 410 | |||
XM_011533161.2 | XP_011531463.1 | 2 | 219 | |||
XM_011533163.2 | XP_011531465.1 | 2 | 219 | |||
XM_017005264.1 | XP_016860753.1 | 3 | 410 | |||
XM_017005265.1 | XP_016860754.1 | 2 | 219 | |||
XM_017005266.1 | XP_016860755.1 | 2 | 219 | |||
GenBank mRNA | AK074418.1 | - | 3 | 289 | ||
AK290298.1 | - | 2 | 223 | |||
AK308037.1 | - | 2 | 199 | |||
BC117345.1 | - | 2 | 183 | 40125896 | ||
BC117347.1 | - | 2 | 183 | 40125898 | ||
BX476842.1 | - | 2 | 236 | |||
BX640793.1 | - | 2 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
