siRNA Details
Gene Symbol: COX6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004373.3 | NP_004364.2 | 3 | 330 | ||
GenBank mRNA | AK312009.1 | - | 3 | 330 | ||
BC007723.2 | - | 1 | 295 | 4300921 | ||
BC070186.1 | - | 1 | 303 | 6654310 | ||
BC107861.1 | - | 1 | 311 | 4664958 | ||
BP378295.1 | - | 3 | 301 | |||
HE647860.1 | - | 3 | 293 | |||
X15341.1 | - | 3 | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
