siRNA Details
Gene Symbol: ENDOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004435.2 | NP_004426.2 | 2 | 759 | ||
XM_011518347.2 | XP_011516649.1 | 2 | 857 | |||
GenBank mRNA | AK130973.1 | - | 2 | 240 | ||
BC004922.2 | - | 2 | 634 | 3604703 | ||
BC016351.1 | - | 2 | 789 | 4080883 | ||
X79444.1 | - | 2 | 740 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
