siRNA Details
Gene Symbol: SETD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286752.1 | NP_001273681.1 | 9 | 1625 | ||
NM_017438.4 | NP_059134.1 | 8 | 1482 | |||
NR_040087.2 | 7 | 1393 | ||||
XM_011529636.1 | XP_011527938.1 | 8 | 1531 | |||
XM_011529637.1 | XP_011527939.1 | 8 | 1596 | |||
XM_011529638.2 | XP_011527940.1 | 8 | 1504 | |||
XM_011529639.1 | XP_011527941.1 | 8 | 1282 | |||
XM_011529640.2 | XP_011527942.1 | 8 | 1547 | |||
XM_011529642.1 | XP_011527944.1 | 9 | 1625 | |||
XM_011529643.1 | XP_011527945.1 | 9 | 1576 | |||
XM_011529644.1 | XP_011527946.1 | 9 | 1137 | |||
XM_017028403.1 | XP_016883892.1 | 8 | 1073 | |||
XM_017028404.1 | XP_016883893.1 | 9 | 1181 | |||
GenBank mRNA | AF391112.1 | - | 7 | 960 | ||
AK001660.1 | - | 8 | 1049 | |||
AK095215.1 | - | 7 | 2349 | |||
AK126368.1 | - | 1 | 1559 | |||
BC036556.2 | - | 9 | 1299 | 5266627 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
