siRNA Details
Gene Symbol: ENDOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011518347.2 | XP_011516649.1 | 1 | 83 | ||
GenBank mRNA | BC016351.1 | - | 15 | 4080883 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
