siRNA Details
Gene Symbol: ARHGAP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199417.1 | NP_001186346.1 | 24 | 3561 | ||
XM_006721991.1 | XP_006722054.1 | 24 | 3602 | |||
XM_006721992.3 | XP_006722055.1 | 24 | 3340 | |||
XM_006721993.3 | XP_006722056.1 | 23 | 3491 | |||
XM_006721994.3 | XP_006722057.1 | 25 | 3610 | |||
XM_011525073.1 | XP_011523375.1 | 24 | 3521 | |||
XM_011525074.2 | XP_011523376.1 | 24 | 3620 | |||
XM_011525075.2 | XP_011523377.1 | 19 | 3162 | |||
GenBank mRNA | AB040934.1 | - | 19 | 2347 | ||
BC039190.1 | - | 18 | 1857 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
