siRNA Details
Gene Symbol: UBE2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256355.1 | NP_001243284.1 | 2 | 269 | ||
NM_003347.3 | NP_003338.1 | 2 | 290 | |||
NR_028436.2 | 2 | 290 | ||||
NR_046082.1 | 2 | 713 | ||||
XM_017028935.1 | XP_016884424.1 | 2 | 167 | |||
GenBank mRNA | AJ000519.1 | - | 2 | 107 | ||
AK125396.1 | - | 2 | 713 | |||
AK291083.1 | - | 2 | 102 | |||
AK299985.1 | - | 2 | 269 | |||
AK311761.1 | - | 2 | 104 | |||
BC053368.1 | - | 2 | 143 | 4214359 | ||
BU567707.1 | - | 2 | 111 | |||
BU584304.1 | - | 2 | 169 | |||
CD690076.1 | - | 2 | 145 | |||
CR456606.1 | - | 2 | 114 | |||
DA271653.1 | - | 2 | 292 | |||
S81003.1 | - | 2 | 107 | |||
X92962.1 | - | 2 | 92 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
