siRNA Details
Gene Symbol: LOC101929479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022812.1 | XP_016878301.1 | 3 | 443 |
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 3 | 470 | |||
NR_033936.3 | 2 | 381 | ||||
NR_103496.2 | 2 | 331 | ||||
GenBank mRNA | AK309420.1 | - | 3 | 421 | ||
AK309732.1 | - | 2 | 332 | |||
AK310769.1 | - | 2 | 284 | |||
AK311124.1 | - | 2 | 236 | |||
BC007811.2 | - | 3 | 394 | |||
BC065260.1 | - | 3 | 387 | |||
BC072390.1 | - | 2 | 334 | |||
DB070142.1 | - | 3 | 470 |
Gene Symbol: GOLGA2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027001.1 | 2 | 364 | |||
GenBank mRNA | AK074778.1 | - | 3 | 421 | ||
BC071951.1 | - | 2 | 321 | |||
BC096759.1 | - | 2 | 321 |
Gene Symbol: LOC642423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK308544.1 | - | 3 | 416 |
Gene Symbol: LOC727751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102747.1 | 2 | 377 | |||
NR_102748.1 | 2 | 327 | ||||
GenBank mRNA | AK291684.1 | - | 3 | 371 | ||
AK307285.1 | - | 2 | 272 | |||
AK309746.1 | - | 2 | 354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
