siRNA Details
Gene Symbol: GOLGA2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026811.3 | 3 | 552 | |||
NR_033936.3 | 2 | 463 | ||||
NR_103496.2 | 2 | 413 | ||||
GenBank mRNA | AK026448.1 | - | 3 | 337 | ||
AK309732.1 | - | 2 | 414 | |||
AK310769.1 | - | 2 | 366 | |||
AK311124.1 | - | 2 | 318 | |||
AK311170.1 | - | 2 | 373 | |||
BC007811.2 | - | 3 | 476 | |||
BC065260.1 | - | 3 | 469 | |||
BC072390.1 | - | 2 | 416 |
Gene Symbol: GOLGA2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027001.1 | 2 | 446 | |||
GenBank mRNA | AK074778.1 | - | 3 | 503 | ||
AK307326.1 | - | 3 | 503 | |||
AK309056.1 | - | 3 | 370 | |||
AK311265.1 | - | 2 | 368 | |||
BC054509.1 | - | 2 | 263 | |||
BC071951.1 | - | 2 | 403 | |||
BC096759.1 | - | 2 | 403 |
Gene Symbol: LOC101929479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022812.1 | XP_016878301.1 | 3 | 525 |
Gene Symbol: LOC727751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102748.1 | 2 | 409 | |||
GenBank mRNA | AK291684.1 | - | 3 | 453 | ||
AK307285.1 | - | 2 | 354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
