siRNA Details
Gene Symbol: LST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166538.1 | NP_001160010.1 | 1 | 13 | ||
NM_007161.3 | NP_009092.3 | 1 | 13 | |||
NM_205837.2 | NP_995309.2 | 2 | 122 | |||
NM_205838.2 | NP_995310.2 | 2 | 150 | |||
NM_205839.2 | NP_995311.2 | 2 | 150 | |||
NM_205840.2 | NP_995312.2 | 1 | 13 | |||
NR_029461.1 | 2 | 150 | ||||
NR_029462.1 | 2 | 150 | ||||
XM_006715206.3 | XP_006715269.1 | 2 | 142 | |||
XM_006715209.3 | XP_006715272.1 | 2 | 138 | |||
XM_006715210.3 | XP_006715273.1 | 2 | 144 | |||
XM_011514914.2 | XP_011513216.1 | 2 | 123 | |||
GenBank mRNA | AF000424.1 | - | 2 | 122 | ||
AF000425.1 | - | 2 | 128 | |||
AF000426.1 | - | 2 | 105 | |||
AJ705986.1 | - | 2 | 87 | |||
AV711621.1 | - | 2 | 114 | |||
AV715235.1 | - | 2 | 205 | |||
AW925228.1 | - | 2 | 150 | |||
BC103855.1 | - | 1 | 18 | 40003000 | ||
BC103856.1 | - | 1 | 18 | 40003004 | ||
BI838080.1 | - | 2 | 108 | |||
BX647293.1 | - | 2 | 135 | |||
Y18486.1 | - | 1 | 13 | |||
Y18487.1 | - | 1 | 13 | |||
Y18488.1 | - | 1 | 13 | |||
Y18489.1 | - | 1 | 13 | |||
Y18490.1 | - | 1 | 13 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
