siRNA Details
Gene Symbol: LST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007161.3 | NP_009092.3 | 1, 2 | 115 | ||
NM_205837.2 | NP_995309.2 | 2, 3 | 224 | |||
NM_205839.2 | NP_995311.2 | 2, 3 | 252 | |||
NM_205840.2 | NP_995312.2 | 1, 2 | 115 | |||
XM_006715206.3 | XP_006715269.1 | 2, 3 | 244 | |||
XM_006715210.3 | XP_006715273.1 | 2, 3 | 246 | |||
XM_011514914.2 | XP_011513216.1 | 2, 3 | 225 | |||
GenBank mRNA | AF000424.1 | - | 2, 3 | 224 | ||
AF000425.1 | - | 2, 3 | 230 | |||
AY852265.1 | - | 1, 2 | 15 | |||
AY852266.1 | - | 1, 2 | 15 | |||
Y18487.1 | - | 1, 2 | 115 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
