siRNA Details
Gene Symbol: ADH1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000669.4 | NP_000660.1 | 3, 4 | 628 | ||
NR_133005.1 | 3, 4 | 628 | ||||
XM_011531588.2 | XP_011529890.1 | 2, 3 | 418 | |||
XM_011531589.1 | XP_011529891.1 | 4, 5 | 748 | |||
GenBank mRNA | BC062476.1 | - | 3, 4 | 324 | 30408761 | |
BC066227.1 | - | 3, 4 | 286 | 7002195 | ||
BC066228.1 | - | 3, 4 | 286 | 7002197 | ||
BC067419.1 | - | 3, 4 | 286 | 7002191 | ||
BC067420.1 | - | 3, 4 | 286 | 7002192 | ||
BC067421.1 | - | 3, 4 | 286 | 7002193 | ||
BC067422.1 | - | 3, 4 | 286 | 7002194 | ||
BC074771.2 | - | 3, 4 | 286 | 30915457 | ||
BC074786.2 | - | 3, 4 | 285 | 30915344 | ||
CB114902.1 | - | 3, 4 | 343 | |||
CD014078.1 | - | 3, 4 | 329 | |||
M12272.1 | - | 3, 4 | 338 | |||
X04299.1 | - | 3, 4 | 330 | |||
X04350.1 | - | 1, 2 | 100 |
Gene Symbol: ADH1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000667.3 | NP_000658.1 | 3, 4 | 372 | ||
GenBank mRNA | AK290558.1 | - | 3, 4 | 329 | ||
AK309597.1 | - | 3, 4 | 347 | |||
BC074738.2 | - | 3, 4 | 286 | 30915195 | ||
BC117442.1 | - | 3, 4 | 328 | 40125993 | ||
BC126306.1 | - | 3, 4 | 328 | 8992022 | ||
BT019812.1 | - | 3, 4 | 258 | |||
M12271.1 | - | 3, 4 | 273 | |||
M12963.1 | - | 3, 4 | 330 |
Gene Symbol: ADH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000668.5 | NP_000659.2 | 3, 4 | 369 | ||
NM_001286650.1 | NP_001273579.1 | 4, 5 | 491 | |||
GenBank mRNA | AF153821.1 | - | 3, 4 | 326 | ||
AK299409.1 | - | 3, 4 | 326 | |||
AK301018.1 | - | 4, 5 | 450 | |||
AK304396.1 | - | 3, 4 | 324 | |||
AK316501.1 | - | 3, 4 | 326 | |||
BC033009.2 | - | 3, 4 | 328 | 4828235 | ||
BP317995.1 | - | 3, 4 | 342 | |||
BX647469.1 | - | 3, 4 | 2353 | |||
D00137.1 | - | 3, 4 | 278 | |||
DC392123.1 | - | 3, 4 | 371 | |||
FJ224285.1 | - | 3, 4 | 328 | |||
M21692.1 | - | 3, 4 | 290 | |||
M24317.1 | - | 3, 4 | 288 | |||
X03350.1 | - | 3, 4 | 330 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
