siRNA Details
Gene Symbol: ADH1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000669.4 | NP_000660.1 | 4 | 662 | ||
NR_133005.1 | 4 | 662 | ||||
XM_011531588.2 | XP_011529890.1 | 3 | 452 | |||
XM_011531589.1 | XP_011529891.1 | 5 | 782 | |||
GenBank mRNA | BC062476.1 | - | 4 | 358 | 30408761 | |
BC066227.1 | - | 4 | 320 | 7002195 | ||
BC066228.1 | - | 4 | 320 | 7002197 | ||
BC067419.1 | - | 4 | 320 | 7002191 | ||
BC067420.1 | - | 4 | 320 | 7002192 | ||
BC067421.1 | - | 4 | 320 | 7002193 | ||
BC067422.1 | - | 4 | 320 | 7002194 | ||
BC074771.2 | - | 4 | 320 | 30915457 | ||
BC074786.2 | - | 4 | 319 | 30915344 | ||
CB114902.1 | - | 4 | 377 | |||
CD014078.1 | - | 4 | 363 | |||
M12272.1 | - | 4 | 372 | |||
X04299.1 | - | 4 | 364 | |||
X04350.1 | - | 2 | 134 |
Gene Symbol: ADH1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000668.5 | NP_000659.2 | 4 | 403 | ||
NM_001286650.1 | NP_001273579.1 | 5 | 525 | |||
GenBank mRNA | AF153821.1 | - | 4 | 360 | ||
AK299409.1 | - | 4 | 360 | |||
AK301018.1 | - | 5 | 484 | |||
AK316501.1 | - | 4, 5 | 360 | |||
BC033009.2 | - | 4 | 362 | 4828235 | ||
BP317995.1 | - | 4 | 376 | |||
BX641105.1 | - | 1 | 2598 | |||
BX647469.1 | - | 4 | 2387 | |||
D00137.1 | - | 4 | 312 | |||
DC392123.1 | - | 4 | 405 | |||
FJ224285.1 | - | 4 | 362 | |||
M21692.1 | - | 4 | 324 | |||
M24317.1 | - | 4 | 322 | |||
X03350.1 | - | 4 | 364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
