siRNA Details
Gene Symbol: LRP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002336.2 | NP_002327.2 | 23 | 6812 | ||
XM_006719078.3 | XP_006719141.1 | 24 | 6274 | |||
XM_011520671.2 | XP_011518973.1 | 22 | 6562 | |||
XM_017019304.1 | XP_016874793.1 | 21 | 6630 | |||
GenBank mRNA | AB209683.1 | - | 24 | 4543 | ||
AK124760.1 | - | 1 | 576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
