siRNA Details
Gene Symbol: NDUFB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278645.1 | NP_001265574.1 | 3 | 398 | ||
NM_001278646.1 | NP_001265575.1 | 3 | 431 | |||
NM_001311168.1 | NP_001298097.1 | 3 | 400 | |||
NM_005005.2 | NP_004996.1 | 3 | 433 | |||
GenBank mRNA | AF044956.1 | - | 3 | 427 | ||
AF067168.1 | - | 3 | 427 | |||
AF261090.1 | - | 3 | 432 | |||
AI907018.1 | - | 3 | 450 | |||
AK026538.1 | - | 3 | 416 | |||
AK302286.1 | - | 3 | 395 | |||
AK313432.1 | - | 3 | 395 | |||
AL136714.1 | - | 3 | 403 | |||
BC007672.2 | - | 3 | 400 | 3160562 | ||
BG703246.1 | - | 3 | 360 | |||
BQ685604.1 | - | 3 | 383 | |||
CN410734.1 | - | 3 | 341 | |||
DB504123.1 | - | 3 | 358 | |||
S82655.1 | - | 2 | 119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
