siRNA Details
Gene Symbol: PMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000535.6 | NP_000526.2 | 3 | 312 | ||
NM_001322003.1 | NP_001308932.1 | 3 | 307 | |||
NM_001322004.1 | NP_001308933.1 | 2 | 172 | |||
NM_001322005.1 | NP_001308934.1 | 3 | 424 | |||
NM_001322006.1 | NP_001308935.1 | 3 | 312 | |||
NM_001322007.1 | NP_001308936.1 | 3 | 312 | |||
NM_001322008.1 | NP_001308937.1 | 2 | 172 | |||
NM_001322009.1 | NP_001308938.1 | 3 | 419 | |||
NM_001322010.1 | NP_001308939.1 | 2 | 172 | |||
NM_001322011.1 | NP_001308940.1 | 3 | 312 | |||
NM_001322012.1 | NP_001308941.1 | 3 | 307 | |||
NM_001322013.1 | NP_001308942.1 | 3 | 307 | |||
NM_001322014.1 | NP_001308943.1 | 3 | 312 | |||
NM_001322015.1 | NP_001308944.1 | 3 | 307 | |||
NR_136154.1 | 3 | 312 | ||||
XM_017012342.1 | XP_016867831.1 | 3 | 286 | |||
GenBank mRNA | AB103082.1 | - | 3 | 225 | ||
AB103083.1 | - | 3 | 225 | |||
AB103085.1 | - | 2 | 225 | |||
AK294661.1 | - | 2 | 162 | |||
AK312390.1 | - | 3 | 261 | |||
BC008400.1 | - | 3 | 270 | |||
BC031832.1 | - | 3 | 276 | |||
BC093921.1 | - | 3 | 247 | 7939766 | ||
BX537558.1 | - | 3 | 426 | |||
CR981250.1 | - | 3 | 213 | |||
DA774159.1 | - | 3 | 312 | |||
U14658.1 | - | 3 | 227 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 8 | 1297 | |||
GenBank mRNA | U38980.1 | - | 8 | 1183 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 3 | 236 |
Gene Symbol: PMS2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003613.1 | 3 | 672 | |||
GenBank mRNA | AB017004.1 | - | 3 | 672 | ||
BC067809.1 | - | 3 | 304 | |||
BC110395.1 | - | 3 | 280 | |||
D38435.1 | - | 3 | 275 | |||
D38499.1 | - | 3 | 275 |
Gene Symbol: PMS2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047753.1 | - | 3 | 232 |
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003614.3 | 3 | 256 | |||
GenBank mRNA | DA215155.1 | - | 3 | 243 |
Gene Symbol: PMS2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028059.1 | 6 | 1058 | |||
GenBank mRNA | BC101598.1 | - | 5 | 509 | 8069104 | |
D38437.1 | - | 2 | 207 | |||
D38498.1 | - | 2 | 207 |
Gene Symbol: PMS2P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_022007.2 | 3 | 269 | |||
NR_046297.1 | 3 | 269 | ||||
NR_046299.1 | 3 | 269 | ||||
GenBank mRNA | BC029368.1 | - | 3 | 246 | ||
BC029419.1 | - | 3 | 234 | |||
CB141754.1 | - | 3 | 234 | |||
D38438.1 | - | 3 | 240 | |||
D38500.1 | - | 3 | 240 | |||
DA885243.1 | - | 3 | 270 |
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 3 | 281 | |||
NR_027776.2 | 3 | 281 | ||||
GenBank mRNA | AK309660.1 | - | 3 | 229 | ||
BF984173.1 | - | 3 | 280 | |||
D38436.1 | - | 3 | 262 | |||
D38439.1 | - | 2 | 193 | |||
D38501.1 | - | 2 | 193 | |||
D38502.1 | - | 3 | 262 | |||
DW407867.1 | - | 3 | 298 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 4 | 434 |
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 3 | 297 | |||
GenBank mRNA | AB017006.1 | - | 3 | 392 | ||
DB472687.1 | - | 3 | 297 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 2 | 672 | |||
GenBank mRNA | BM845848.1 | - | 2 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
