siRNA Details
Gene Symbol: C21orf91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100421.1 | NP_001093891.1 | 3, 4 | 759 | ||
GenBank mRNA | AF239726.1 | - | 3, 4 | 712 | ||
AK225123.1 | - | 3, 4 | 730 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
