siRNA Details
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 3 | 256 | |||
NR_027776.2 | 3 | 256 | ||||
GenBank mRNA | AK309660.1 | - | 3 | 204 | ||
BF984173.1 | - | 3 | 255 | |||
D38436.1 | - | 3 | 237 | |||
D38439.1 | - | 2 | 168 | |||
D38501.1 | - | 2 | 168 | |||
D38502.1 | - | 3 | 237 | |||
DW407867.1 | - | 3 | 273 |
Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023383.1 | 8 | 1272 | |||
GenBank mRNA | U38980.1 | - | 8 | 1158 |
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 3 | 211 |
Gene Symbol: PMS2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003613.1 | 3 | 647 | |||
GenBank mRNA | AB017004.1 | - | 3 | 647 | ||
BC067809.1 | - | 3 | 279 | |||
BC110395.1 | - | 3 | 255 | |||
D38435.1 | - | 3 | 250 | |||
D38499.1 | - | 3 | 250 |
Gene Symbol: PMS2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047753.1 | - | 3 | 207 |
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003614.3 | 3 | 231 | |||
GenBank mRNA | DA215155.1 | - | 3 | 218 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 4 | 409 |
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 3 | 272 | |||
GenBank mRNA | AB017006.1 | - | 3 | 367 | ||
DB472687.1 | - | 3 | 272 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028058.1 | 2 | 647 | |||
GenBank mRNA | BM845848.1 | - | 2 | 75 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
