siRNA Details
Gene Symbol: MST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020998.3 | NP_066278.3 | 18 | 2289 | ||
XM_006713166.1 | XP_006713229.1 | 17 | 2919 | |||
XM_011533732.1 | XP_011532034.1 | 18 | 3090 | |||
XM_017006460.1 | XP_016861949.1 | 18 | 3033 | |||
XM_017006461.1 | XP_016861950.1 | 18 | 2997 | |||
XM_017006462.1 | XP_016861951.1 | 18 | 3083 | |||
XM_017006463.1 | XP_016861952.1 | 18 | 2978 | |||
XM_017006464.1 | XP_016861953.1 | 18 | 2921 | |||
GenBank mRNA | AK222893.1 | - | 18 | 2180 | ||
BC048330.1 | - | 18 | 2188 | 5190966 | ||
L11924.1 | - | 18 | 2185 | |||
M74178.1 | - | 18 | 2175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
