siRNA Details
Gene Symbol: ANKRD36BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015424.1 | 9 | 819 | |||
GenBank mRNA | BC031698.1 | - | 9 | 819 | ||
BX648045.1 | - | 9 | 853 |
Gene Symbol: ANKRD36C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310154.1 | NP_001297083.1 | 30 | 2593 | ||
GenBank mRNA | AK123626.1 | - | 24 | 1865 | ||
AL832836.1 | - | 11 | 493 | |||
AL832977.1 | - | 3 | 58 | |||
U28831.1 | - | 11 | 521 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
