siRNA Details
Gene Symbol: PHOSPHO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143804.1 | NP_001137276.1 | 1 | 101 | ||
NM_178500.3 | NP_848595.1 | 1 | 101 | |||
GenBank mRNA | AJ457189.1 | - | 1 | 101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
