siRNA Details
Gene Symbol: RRN3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003370.2 | 6 | 496 | |||
GenBank mRNA | AL702574.1 | - | 1 | 77 | ||
BC009198.2 | - | 7 | 726 | |||
BC068999.1 | - | 7 | 726 | |||
BX101439.1 | - | 6 | 586 | |||
CN397262.1 | - | 6 | 503 |
Gene Symbol: RRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301064.1 | NP_001287993.1 | 5 | 531 | ||
NM_018427.4 | NP_060897.3 | 6 | 621 | |||
XM_005255376.1 | XP_005255433.1 | 6 | 530 | |||
XM_005255377.4 | XP_005255434.1 | 5 | 479 | |||
XM_011522541.1 | XP_011520843.1 | 6 | 530 | |||
GenBank mRNA | AF227156.1 | - | 6 | 587 | ||
AJ272050.1 | - | 6 | 541 | |||
AK225662.1 | - | 6 | 597 | |||
AK295486.1 | - | 3 | 214 | |||
AK300253.1 | - | 5 | 440 | |||
AK302994.1 | - | 3 | 268 | |||
AK303405.1 | - | 3 | 213 | |||
AK304856.1 | - | 6 | 487 | |||
AK314769.1 | - | 6 | 535 | |||
AK316520.1 | - | 5 | 445 | |||
BC036182.1 | - | 6 | 536 | |||
BC104660.1 | - | 6 | 543 | |||
BC132688.1 | - | 6 | 538 | 40146710 | ||
BC132690.1 | - | 6 | 538 | 40146712 |
Gene Symbol: RRN3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003369.2 | 6 | 710 | |||
GenBank mRNA | AK225670.1 | - | 6 | 558 | ||
AK298442.1 | - | 5 | 443 | |||
AK301253.1 | - | 6 | 530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
