siRNA Details
Gene Symbol: RRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301064.1 | NP_001287993.1 | 5, 6 | 554 | ||
NM_018427.4 | NP_060897.3 | 6, 7 | 644 | |||
XM_011522541.1 | XP_011520843.1 | 6, 7 | 553 | |||
GenBank mRNA | AF227156.1 | - | 6, 7 | 610 | ||
AJ272050.1 | - | 6, 7 | 564 | |||
AK225662.1 | - | 6, 7 | 620 | |||
AK295486.1 | - | 3, 4 | 237 | |||
AK300253.1 | - | 5, 6 | 463 | |||
AK302994.1 | - | 3, 4 | 291 | |||
AK303405.1 | - | 3, 4 | 236 | |||
AK304856.1 | - | 6, 7 | 510 | |||
AK314769.1 | - | 6, 7 | 558 | |||
AK316520.1 | - | 5, 6 | 468 | |||
BC036182.1 | - | 6, 7 | 559 | |||
BC104660.1 | - | 6, 7 | 566 | |||
BC132688.1 | - | 6, 7 | 561 | 40146710 | ||
BC132690.1 | - | 6, 7 | 561 | 40146712 |
Gene Symbol: RRN3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL702574.1 | - | 1, 2 | 100 | ||
BC009198.2 | - | 7, 8 | 749 | |||
BC068999.1 | - | 7, 8 | 749 | |||
BX101439.1 | - | 6, 7 | 609 |
Gene Symbol: RRN3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003369.2 | 6, 7 | 733 | |||
GenBank mRNA | AK225670.1 | - | 6, 7 | 581 | ||
AK298442.1 | - | 5, 6 | 466 | |||
AK301253.1 | - | 6, 7 | 553 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
