siRNA Details
Gene Symbol: RRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301064.1 | NP_001287993.1 | 7, 8 | 684 | ||
NM_018427.4 | NP_060897.3 | 8, 9 | 774 | |||
XM_011522540.2 | XP_011520842.1 | 3, 4 | 186 | |||
GenBank mRNA | AF227156.1 | - | 8, 9 | 740 | ||
AJ272050.1 | - | 8, 9 | 694 | |||
AK225662.1 | - | 8, 9 | 750 | |||
AK295486.1 | - | 5, 6 | 367 | |||
AK300253.1 | - | 7, 8 | 593 | |||
AK302994.1 | - | 5, 6 | 421 | |||
AK304856.1 | - | 8, 9 | 640 | |||
AK314769.1 | - | 8, 9 | 688 | |||
AK316520.1 | - | 7, 8 | 598 | |||
BC036182.1 | - | 8, 9 | 689 | |||
BC104660.1 | - | 8, 9 | 696 | |||
BC132688.1 | - | 8, 9 | 691 | 40146710 | ||
BC132690.1 | - | 8, 9 | 691 | 40146712 |
Gene Symbol: RRN3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003369.2 | 8, 9 | 861 | |||
GenBank mRNA | AK225670.1 | - | 8, 9 | 711 | ||
AK301253.1 | - | 8, 9 | 681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
