siRNA Details
Gene Symbol: UQCRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297565.1 | NP_001284494.1 | 5 | 751 | ||
NM_001297566.1 | NP_001284495.1 | 3 | 576 | |||
NM_006004.3 | NP_005995.2 | 4 | 603 | |||
GenBank mRNA | BC001426.2 | - | 4 | 503 | 3140323 | |
BC001934.1 | - | 4 | 508 | 2823237 | ||
BC015177.1 | - | 4 | 506 | 3905169 | ||
BC093060.1 | - | 5 | 666 | 30409772 | ||
BC107703.1 | - | 4 | 509 | 6605146 | ||
M36647.1 | - | 4 | 482 | |||
Y00764.1 | - | 4 | 488 |
Gene Symbol: VGLL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096878.1 | - | 1145 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
