siRNA Details
Gene Symbol: HTR1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000864.4 | NP_000855.1 | 1 | 1012 | ||
GenBank mRNA | AF498979.1 | - | 1 | 502 | ||
BC007720.2 | - | 1 | 1012 | 4299633 | ||
BT007027.1 | - | 1 | 502 | |||
M81589.1 | - | 1 | 545 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
