siRNA Details
Gene Symbol: GPHB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145171.3 | NP_660154.3 | 3 | 308 | ||
GenBank mRNA | AF403430.1 | - | 3 | 251 | ||
AF467770.1 | - | 2 | 251 | |||
BC069113.1 | - | 3 | 251 | 7216930 | ||
BC133702.1 | - | 2 | 251 | |||
BM709443.1 | - | 3 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
