siRNA Details
Gene Symbol: OPLAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017570.4 | NP_060040.1 | 14, 15 | 2092 | ||
XM_011516960.1 | XP_011515262.1 | 14, 15 | 2368 | |||
GenBank mRNA | AB122018.1 | - | 13, 14 | 1988 | ||
AL096750.1 | - | 6, 7 | 1077 | |||
BC036944.1 | - | 9, 10 | 1279 | |||
BC142672.1 | - | 14, 15 | 2092 | 40148181 | ||
BC150206.1 | - | 14, 15 | 2070 | 40148883 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
