siRNA Details
Gene Symbol: OPLAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017570.4 | NP_060040.1 | 24 | 3523 | ||
XM_011516960.1 | XP_011515262.1 | 24 | 3799 | |||
GenBank mRNA | AB122018.1 | - | 23 | 3419 | ||
AF217994.2 | - | 1385 | ||||
AL096750.1 | - | 16 | 2508 | |||
BC036944.1 | - | 19 | 2710 | |||
BC041576.1 | - | 9 | 1334 | |||
BC142672.1 | - | 24 | 3523 | 40148181 | ||
BC150206.1 | - | 24 | 3501 | 40148883 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
