siRNA Details
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102445.2 | NP_001095915.1 | 2 | 507 | ||
NM_001113381.1 | NP_001106852.1 | 1 | 118 | |||
NM_005613.5 | NP_005604.1 | 1 | 398 | |||
GenBank mRNA | AK093959.1 | - | 11 | |||
AK295540.1 | - | 2 | 242 | |||
AK300832.1 | - | 12 | ||||
BI547160.1 | - | 1 | 112 | |||
DA253378.1 | - | 2 | 242 | |||
DA660018.1 | - | 1 | 167 | |||
DA660695.1 | - | 1 | 154 | |||
DC318279.1 | - | 1 | 398 | |||
DC334933.1 | - | 2 | 507 | |||
DQ346662.1 | - | 11 | ||||
DQ346663.1 | - | 1 | 226 | |||
DQ346664.1 | - | 2 | 338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
