siRNA Details
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102445.2 | NP_001095915.1 | 3 | 721 | ||
NM_001113380.1 | NP_001106851.1 | 2 | 296 | |||
NM_001113381.1 | NP_001106852.1 | 2 | 332 | |||
NM_005613.5 | NP_005604.1 | 2 | 612 | |||
GenBank mRNA | AB209019.1 | - | 1 | 1096 | ||
AF493928.1 | - | 2 | 101 | |||
AK093959.1 | - | 2 | 225 | |||
AK096204.1 | - | 2 | 223 | |||
AK295540.1 | - | 3 | 456 | |||
AK296240.1 | - | 2 | 140 | |||
AK300832.1 | - | 2 | 226 | |||
AK312250.1 | - | 2 | 224 | |||
BC000737.1 | - | 2 | 186 | 3510260 | ||
BC051869.1 | - | 2 | 198 | 5786695 | ||
BI547160.1 | - | 2 | 326 | |||
BT007025.1 | - | 2 | 101 | |||
DA253378.1 | - | 3 | 456 | |||
DA660018.1 | - | 2 | 381 | |||
DA660695.1 | - | 2 | 368 | |||
DQ346662.1 | - | 2 | 225 | |||
DQ346663.1 | - | 2 | 440 | |||
DQ346664.1 | - | 3 | 552 | |||
EF054877.1 | - | 2 | 296 | |||
U27768.1 | - | 2 | 198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
