siRNA Details
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102445.2 | NP_001095915.1 | 3 | 745 | ||
NM_001113380.1 | NP_001106851.1 | 2 | 320 | |||
NM_001113381.1 | NP_001106852.1 | 2 | 356 | |||
NM_005613.5 | NP_005604.1 | 2 | 636 | |||
GenBank mRNA | AB209019.1 | - | 1 | 1120 | ||
AF493928.1 | - | 2 | 125 | |||
AK093959.1 | - | 2 | 249 | |||
AK096204.1 | - | 2 | 247 | |||
AK295540.1 | - | 3 | 480 | |||
AK296240.1 | - | 2 | 164 | |||
AK300832.1 | - | 2 | 250 | |||
AK312250.1 | - | 2 | 248 | |||
BC000737.1 | - | 2 | 210 | 3510260 | ||
BC051869.1 | - | 2 | 222 | 5786695 | ||
BI547160.1 | - | 2 | 350 | |||
BT007025.1 | - | 2 | 125 | |||
DA253378.1 | - | 3 | 480 | |||
DA660018.1 | - | 2 | 405 | |||
DA660695.1 | - | 2 | 392 | |||
DQ346662.1 | - | 2 | 249 | |||
DQ346663.1 | - | 2 | 464 | |||
DQ346664.1 | - | 3 | 576 | |||
EF054877.1 | - | 2 | 320 | |||
U27768.1 | - | 2 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
