siRNA Details
Gene Symbol: SPAG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_058203.2 | NP_478110.1 | 3 | 445 | ||
GenBank mRNA | AJ628017.1 | - | 3 | 283 | ||
AJ635328.1 | - | 3 | 435 |
Gene Symbol: SPAG11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005272392.4 | XP_005272449.2 | 3 | 418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
